missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Penicillin-Streptomycin (5000 IU/mL, 5 mg/mL)

Catalog No. MP091670049
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP091670049 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP091670049 Supplier MP Biomedicals™ Supplier No. 091670049

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Dual antibiotic solution. Concentration: Penicillin 5000 IU/mL, Streptomycin 5 mg/mL

The antibiotics penicillin and streptomycin are used to prevent bacterial contamination of cell cultures due to their effective combined action against gram-positive and gram-negative bacteria.

Specifications

Chemical Name or Material Penicillin-Streptomycin
Quantity 100 mL

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.