missing translation for 'onlineSavingsMsg'
Learn More

Penicillin G sodium salt

Catalog No. AAJ6303222
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6303222 100 g
AAJ6303206 5 g
AAJ6303214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6303222 Supplier Thermo Scientific Chemicals Supplier No. J6303222

Penicillin G sodium salt, CAS # 69-57-8, also called benzylpenicillin, is a β-lactam antibiotic active against gram-positive and gram-negative bacteria. | CAS: 69-57-8 | C16H17N2NaO4S | 356.372 g/mol

Penicillin G sodium salt is used as a β-lactam antibiotic. Penicillin G can be used as a selective agent in several types of isolation media. It has been used to study the diagnostic and therapeutic implications of gentamicin-resistant Enterococcus faecalis sequence type 6 with reduced penicillin susceptibility and in cell culture both alone and combined with streptomycin and other antibiotics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Penicillin G sodium salt is the sodium salt of benzylpenicillin, a broad-spectrum β-lactam antibiotic with gram-positive and gram-negative bactericidal activity
  • Penicillin G binds and inactivates penicillin-binding-proteins associated with the bacterial cell wall, thus inhibiting cell wall synthesis and resulting in cell lysis

Applications

  • Penicillin G can be used as a selection agent in the isolation of microorganisms or in cell culture applications
  • It can also be used to prevent infection in research animals

Chemical Identifiers

CAS 69-57-8
Molecular Formula C16H17N2NaO4S
Molecular Weight (g/mol) 356.372
MDL Number MFCD00069666
InChI Key FCPVYOBCFFNJFS-LQDWTQKMSA-M
Synonym Benzylpenicillin sodium salt
PubChem CID 23668834
ChEBI CHEBI:51765
IUPAC Name sodium;(2S,5R,6R)-3,3-dimethyl-7-oxo-6-[(2-phenylacetyl)amino]-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
SMILES CC1(C(N2C(S1)C(C2=O)NC(=O)CC3=CC=CC=C3)C(=O)[O-])C.[Na+]

Specifications

Type Penicillin G Sodium Salt
Physical Form Powder
Color White
Quantity 100 g
Beilstein 3834217
Merck Index 14,7094
Solubility Information Soluble in water (100mg/ml),alcohol,glycerol,DMSO (71mg/ml at 25°C),and ethanol (< 1mg/ml at 25°C).
Formula Weight 356.37
Chemical Name or Material Penicillin G sodium salt
Hazard Category H317-H334
Hazard Statement GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
Precautionary Statement P261-P272-P280g-P285-P302+P352-P304+P341-P333+P313-P342+P311-P363-P501c
EINECSNumber 200-710-2
RTECSNumber XH9800000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only