Learn More
Penicillin G sodium salt
Penicillin G sodium salt, CAS # 69-57-8, also called benzylpenicillin, is a β-lactam antibiotic active against gram-positive and gram-negative bacteria. | CAS: 69-57-8 | C16H17N2NaO4S | 356.372 g/mol
Supplier: Thermo Scientific Chemicals J6303214
| Quantity | 25 g |
|---|
Description
Penicillin G sodium salt is used as a β-lactam antibiotic. Penicillin G can be used as a selective agent in several types of isolation media. It has been used to study the diagnostic and therapeutic implications of gentamicin-resistant Enterococcus faecalis sequence type 6 with reduced penicillin susceptibility and in cell culture both alone and combined with streptomycin and other antibiotics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
- Penicillin G sodium salt is the sodium salt of benzylpenicillin, a broad-spectrum β-lactam antibiotic with gram-positive and gram-negative bactericidal activity
- Penicillin G binds and inactivates penicillin-binding-proteins associated with the bacterial cell wall, thus inhibiting cell wall synthesis and resulting in cell lysis
Applications
- Penicillin G can be used as a selection agent in the isolation of microorganisms or in cell culture applications
- It can also be used to prevent infection in research animals
Chemical Identifiers
| 69-57-8 | |
| 356.372 | |
| FCPVYOBCFFNJFS-LQDWTQKMSA-M | |
| 23668834 | |
| sodium;(2S,5R,6R)-3,3-dimethyl-7-oxo-6-[(2-phenylacetyl)amino]-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate |
| C16H17N2NaO4S | |
| MFCD00069666 | |
| Benzylpenicillin sodium salt | |
| CHEBI:51765 | |
| CC1(C(N2C(S1)C(C2=O)NC(=O)CC3=CC=CC=C3)C(=O)[O-])C.[Na+] |
Specifications
| Penicillin G Sodium Salt | |
| Powder | |
| White | |
| C16H17N2NaO4S | |
| 3834217 | |
| Benzylpenicillin sodium salt | |
| FCPVYOBCFFNJFS-LQDWTQKMSA-M | |
| sodium;(2S,5R,6R)-3,3-dimethyl-7-oxo-6-[(2-phenylacetyl)amino]-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate | |
| 23668834 | |
| 356.37 |
| Penicillin G sodium salt | |
| 69-57-8 | |
| 25 g | |
| MFCD00069666 | |
| 14,7094 | |
| Soluble in water (100mg/ml),alcohol,glycerol,DMSO (71mg/ml at 25°C),and ethanol (< 1mg/ml at 25°C). | |
| CC1(C(N2C(S1)C(C2=O)NC(=O)CC3=CC=CC=C3)C(=O)[O-])C.[Na+] | |
| 356.372 | |
| CHEBI:51765 |
Safety and Handling
GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
P261-P272-P280g-P285-P302+P352-P304+P341-P333+P313-P342+P311-P363-P501c
H317-H334
EINECSNumber : 200-710-2
RTECSNumber : XH9800000
TSCA : No
Recommended Storage : Keep cold