missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ PEN Membrane Glass Slides

Catalog No. 361014590
Change view
Click to view available options
Quantity:
50 each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
361014590 50 each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 361014590 Supplier Applied Biosystems™ Supplier No. LCM0522

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

ARCTURUS™ laser capture microdissection (LCM) instruments offer a unique combination of infrared (IR) laser capture and ultraviolet (UV) laser cutting, permitting the use of any slide type and any sample preparation for microdissection.

ARCTURUS™ laser capture microdissection (LCM) instruments offer a unique combination of infrared (IR) laser capture and ultraviolet (UV) laser cutting, permitting the use of any slide type and any sample preparation for microdissection. While plain glass slides are effectively used for standard IR-based LCM, polyethylene naphthalate (PEN) membrane slides facilitate this Arcturus™-exclusive dual laser function and allow you to determine the appropriate sample preparation based on your application need.

Plain Glass Slides
The ARCTURUS™ instruments are the only LCM systems that allow efficient use of low cost plain glass slides, and provide easy access to archival materials. We do not sell plain glass slides, though most commercially available glass slides are compatible with microdissection. For recommendations to suit your tissue type and application needs, please contact Technical Support for more information.

PEN Membrane Slides
PEN membrane slides enable the combined use of IR LCM and UV Laser Cutting. PEN Membrane Glass Slides allows the use of standard IR LCM on its own, which is the gentlest approach for isolating individual cells or small areas. PEN Membrane Frame Slides provide additional flexibility by allowing the choice of microdissection from non-dehydrated sample preparations.

For Research Use Only. Not for use in diagnostics procedures.

Specifications

Type Membrane Glass Slide
Content And Storage PEN Membrane Glass Slides. Store at room temperature
For Use With (Equipment) ArcturusXT™ LCM Instrument
Shipping Condition Room Temperature
Material PEN Membrane Glass
Quantity 50 each

For Research Use Only. Not for use in diagnostic procedures.