missing translation for 'onlineSavingsMsg'
Learn More

Pemirolast Potassium 98.0+%, TCI America™

Catalog No. p-7135597
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P25471G 1 g
P25475G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P25471G Supplier TCI America Supplier No. P25471G
Only null left

Chemical Identifiers

CAS 100299-08-9
Molecular Formula C10H7KN6O
Molecular Weight (g/mol) 266.305
MDL Number MFCD01690051
InChI Key NMMVKSMGBDRONO-UHFFFAOYSA-N
Synonym 9-Methyl-3-(1H-tetrazol-5-yl)-4H-pyrido[1,2-a]pyrimidin-4-one Potassium Salt
PubChem CID 443866
IUPAC Name potassium;9-methyl-3-(1,2,3-triaza-4-azanidacyclopenta-2,5-dien-5-yl)pyrido[1,2-a]pyrimidin-4-one
SMILES CC1=CC=CN2C1=NC=C(C2=O)C3=NN=N[N-]3.[K+]

Specifications

Melting Point 317°C
Quantity 1 g
Formula Weight 266.31
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Pemirolast Potassium
RTECSNumber UV1164470
TSCA No
Recommended Storage Refrigerator