missing translation for 'onlineSavingsMsg'
Learn More

3M™ PELTOR™ X4 Series Ear Muffs

Catalog No. p-7214256
Change view
Click to view available options
Model:
X4P5E
CSA Z94.2:
Class A
Dielectric:
Yes
Style:
Hard Hat
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model CSA Z94.2 Dielectric Style
19153676 X4P5E Class A Yes Hard Hat
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19153676 Supplier 3M™ Supplier No. 7100097429
Only null left

PELTOR™ X4 Series Ear Muffs combines high attenuation with a sleek, low-profile and lightweight design. Available in over-the-head or hard hat-attached styles.

  • Light and small cup profile with high level of attenuation
  • High level of protection without all the bulk
  • Innovative foams and proprietary high density offers slow recovery
  • Bright yellow-green color cups help employers verify hearing protectors are being worn
  • Available in electrically insulated models (exception is model X4P3E)
  • Compatibility with hard hats (exception is model X4A)

Compliance

3M™ ear muffs must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.

Specifications

Color Black, Chartreuse
ANSI S3.1974 Yes
Cushion Material Foam
CSA Z94.2 Class A
Cushion Color Black
For Use With (Equipment) Fits a wide range of industrial safety hard hats
For Use With (Application) Utilities, Mining, Electrical Contractors, Oil and Gas
Model X4P5E
Pairs per Package 1 Ea.
Noise Reduction Rating (NRR) 25 dB
Product Type X4 Series
Dielectric Yes
Material Foam, Plastic, Metal
Style Hard Hat
Show More Show Less