missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ PELTOR™ ComTac™ V Hearing Defender Headset
PELTOR™ ComTac™ V protects users from hazardous steady-state and impulse noise featuring a more natural sounding headset with excellent sound clarity.
$900.03 - $900.03
Specifications
| ANSI S3.1974 | Yes |
|---|---|
| Cushion Color | Black |
| For Use With (Application) | Law Enforcement, Armed Forces |
| Pairs per Package | 1 Ea. |
| Dielectric | No |
| Catalog Number | Mfr. No. | Model | Color | Foldable | Style | Price | Quantity | ||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Model | Color | Foldable | Style | Price | Quantity | ||
|
19-072-270
|
3M™
7100223427 |
MT20H682FB-09 CY | Coyote Brown | Yes | Over-the-head |
Each for $900.03
Case of 10 Each for $9,000.30
|
|
||
Description
- Compatible with most helmets
- Dynamic Differential Microphone
- Level Dependent Function
- Noise canceling microphone
- Two-Way Communication
- Available in over-the-head or neckband styles
- Choose from three colors: Black, Olive Drab (O.D.) Green or Coyote Brown
Compliance
3M™ ear muffs must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Specifications
| Yes | |
| Law Enforcement, Armed Forces | |
| No |
| Black | |
| 1 Ea. |