Learn More
Thermo Scientific™ Peltier Accessory (Air-Cooled) for GENESYS™ 10S series, BioMate™ 3S and Evolution™ 60S spectrophotometers
Catalog No. 14380006
Change view
Click to view available options
Type:
Cuvette Stir Bars
Catalog No. Type
14380006 Cuvette Stir Bars
1 options
Catalog No. 14380006 Supplier Thermo Scientific™ Supplier No. 840262800
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Thermo Scientific™ Air-Cooled Peltier Accessory for GENESYS 10S series, BioMate 3S and Evolution 60S spectrophotometers quickly specifies the temperature for the analysis and controls the rate of stirring with the external control module.
Thermo Scientific™ Peltier Accessory (Air-Cooled) for GENESYS™ 10S series, BioMate™ 3S and Evolution™ 60S Spectrophotometers quickly specifies the temperature for the analysis and controls the rate of stirring with the external control module. Temperature control and sample stirring are important for many UV-Visible analysis methods. The air-cooled Peltier accessory delivers superior performance in an easy-to-use configuration. It provides reliable temperature control from 20° to 60°C.
The Peltier control system provided high accuracy temperature control without the need to watch water levels, mix in anti-freeze or anti-bacterial, or plumb the accessory with hoses and clamps.
Control module features a large temperature display and graphical indication of the stirring speed
Module software allows you to change temperature or stirring speed quickly
Offers exceptional temperature stability and fast temperature transitions
Small temperature differential between set temperature of the accessory and temperature as measured inside a standard, 1cm cuvette
Less expensive than most recirculating liquid temperature controllers
No maintenance design
Specifications
Type Cuvette Stir Bars
For Use With (Equipment) Peltier thermostatted cell holders