missing translation for 'onlineSavingsMsg'
Learn More

Pelitinib 97.0+%, TCI America™

Supplier:  TCI America P2529100MG

 View more versions of this product

Catalog No. P2529100MG


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Pelitinib
198°C
C24H23ClFN5O2
100 mg
EKB 569, (2E)-N-[4-[(3-Chloro-4-fluorophenyl)amino]-3-cyano-7-ethoxy-6-quinolinyl]-4-dimethylamino-2-butenamide
CCOC1=C(C=C2C(=C1)N=CC(=C2NC3=CC(=C(C=C3)F)Cl)C#N)NC(=O)C=CCN(C)C
467.929
CHEBI:38927
≥97.0% (HPLC)
257933-82-7
White-Yellow
MFCD09837868
2811
WVUNYSQLFKLYNI-AATRIKPKSA-N
(E)-N-[4-(3-chloro-4-fluoroanilino)-3-cyano-7-ethoxyquinolin-6-yl]-4-(dimethylamino)but-2-enamide
6445562
467.93
Crystalline Powder

Chemical Identifiers

257933-82-7
467.929
WVUNYSQLFKLYNI-AATRIKPKSA-N
6445562
(E)-N-[4-(3-chloro-4-fluoroanilino)-3-cyano-7-ethoxyquinolin-6-yl]-4-(dimethylamino)but-2-enamide
C24H23ClFN5O2
MFCD09837868
EKB 569, (2E)-N-[4-[(3-Chloro-4-fluorophenyl)amino]-3-cyano-7-ethoxy-6-quinolinyl]-4-dimethylamino-2-butenamide
CHEBI:38927
CCOC1=C(C=C2C(=C1)N=CC(=C2NC3=CC(=C(C=C3)F)Cl)C#N)NC(=O)C=CCN(C)C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Freezer

SDS
Documents

Documents

Promotions

Promotions