missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pelican™ SabreLite™ 2010 Recoil LED™ Flashlights
Description
- 32 hours of burn time
- EXL body
- Twist ON/OFF lens shroud eliminates accidental discharge
Order Info
Length: 8.1 in. (20.6cm). Weight (with batteries): 13.4 oz. (380g). Requires three C alkaline batteries.
Certifications
ETL listed for certain Class I, II, III, Division 1 hazardous locations. 1ECEx marked.
Specifications
Specifications
| Light Source | LED |
| Lighting Life | 10,000 hr. |
| Voltage | 4.5 V |
| Weight (English) | 13.4 oz. |
| Weight (Metric) | 0.38 kg |
| Battery Run Time | 32 hr. |
| Battery Type | Alkaline |
| Certifications/Compliance | ETL, Class I, II, III, division 1, 1ECEx |
| Color | Black |
| Controller Type | On/Off |
| Show More |