missing translation for 'onlineSavingsMsg'
Learn More

Pelican™ SabreLite™ 2010 Recoil LED™ Flashlights

Catalog No. 19412642
Encompass
Change view
Click to view available options
Product Type:
Flashlight
Includes:
Three C alkaline batteries
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Includes
19412642 Flashlight Three C alkaline batteries
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19412642 Supplier Pelican™ Supplier No. 2010 010 110

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Produce a white collimated beam as bright as incandescent light

  • 32 hours of burn time
  • EXL body
  • Twist ON/OFF lens shroud eliminates accidental discharge

Order Info

Length: 8.1 in. (20.6cm). Weight (with batteries): 13.4 oz. (380g). Requires three C alkaline batteries.

Certifications

ETL listed for certain Class I, II, III, Division 1 hazardous locations. 1ECEx marked.

Specifications

Light Source LED
Lighting Life 10,000 hr.
Voltage 4.5 V
Weight (English) 13.4 oz.
Weight (Metric) 0.38 kg
Battery Run Time 32 hr.
Battery Type Alkaline
Certifications/Compliance ETL, Class I, II, III, division 1, 1ECEx
Color Black
Controller Type On/Off
For Use With (Application) Produce a white collimated beam as bright as incandescent light
Includes Three C alkaline batteries
Length (English) 8.11 in.
Length (Metric) 20.6 cm
Lumen 109 lm
Material ABS Resin
Product Type Flashlight
Show More Show Less