missing translation for 'onlineSavingsMsg'
Learn More

Pelican™ HeadsUp™ Lites Headlamps

Hands-free light when and where you need it

$82.65

Specifications

Battery Type Alkaline
Certifications/Compliance UL, Class I, Division 2, CE
Color Black
For Use With (Application) Hands free light when and where you need it, adjustable cloth strap fits comfortably and securely on head
Material ABS Resin
View More Specs

Products 1
Catalog Number Mfr. No. Battery Run Time Includes Length (Metric) Lumen Price Quantity & Availability  
Catalog Number Mfr. No. Battery Run Time Includes Length (Metric) Lumen Price Quantity & Availability  
19130091
View Documents Promotion Details
Pelican™
2610
19 hr. 3 AAA alkaline batteries, adjustable cloth strap and rubber strap 3.8 cm 30 lm
Each for $82.65
Only null left
 
Description

Description

  • Adjustable cloth strap fits comfortably and securely on head
  • Rubber strap grips tightly to hardhats and helmets

2610 Model

  • 47-hour burn time on low; 19 hours on high
  • Dual-mode button activates one or all three LEDs for full brightness
  • Length: 1.5 in. (3.8cm)
  • Weight (with batteries): 3.2 oz. (90.1g)
  • Powered by three AAA alkaline cells (included)

2620 Model

  • 18-hour burn time on low; 2.25 hours on high
  • Hybrid dual lamps; Xenon for intense bright light or three LEDs
  • Multi-angle pivoting head allows light to be aimed where needed
  • Length: 2.2 in. (5.5cm)
  • Weight (with batteries): 4.8 oz. (136kg)
  • Powered by three AAA alkaline cells (included)

2680 Model

  • Outfitted with Recoil LED module
  • Uses a 1 watt LED aimed backwards into the reflector
  • 100% of the light is captured and aimed forward as a tight collimated beam as bright as an incandescent
  • Body is made of tough, corrosion proof, submersible, and virtually indestructible ABS resin
  • Powered by four AA alkaline cells (included)
  • Comes with a rubber strap for hard hat use and an adjustable cloth strap for bare head

2690 Model

  • Impact and chemical-resistant ABS'water resistant for all-weather use
  • Provides 60 lumens of LED light
  • 10 hours of battery burn time
  • With rotary bezel switch
  • Adjustable cloth strap; heavy-duty rubber strap
  • Powered by three AAA alkaline cells (included)

Certifications

2610, 2680, and 2690 Models are UL listed for certain Class I, Division 1 hazardous locations; CE marked. 2680 Model is UL listed for certain Class I, Division 2 hazardous locations; CE marked.

Specifications

Specifications

Alkaline
Black
ABS Resin
Headlamp
UL, Class I, Division 2, CE
Hands free light when and where you need it, adjustable cloth strap fits comfortably and securely on head
Lifetime
Videos
SDS
Documents

Documents