missing translation for 'onlineSavingsMsg'
Learn More

Pelican™ 9430 Remote Area Lighting System

Catalog No. 19819173
Encompass
Change view
Click to view available options
Light Source:
High Flux LED
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Light Source
19819173 High Flux LED
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19819173 Supplier Pelican™ Supplier No. 0943000001245

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Provides up to 15 hrs of run time with 3000 lumens of brightness

  • Maintenance free LED array with life expectancy of 50,000 hours
  • Fully extendable mast with 360° rotating head
  • Low battery warning system
  • Waterproof rubber activation switch
  • Mast extension lock
  • Switch type: Silicone rubber keypad
  • 12v Rechargeable, sealed battery
  • Battery life: 500 cycles
  • LED type: High Flux LED
  • Lumen value: 3000 (High)/1500 (Low)
  • Voltage: 12V
  • Weight: 22.65 lb. (10.27kg)
  • L x W x D: 6.5 x 7.87 x 2.56 in. (16.5 x 20 x 6.5cm)

Certifications

RoHS (Restriction of Hazardous Substances).

Compliance

CE.

Specifications

Light Source High Flux LED
Voltage 12 V
Battery Life 500 cycles
Battery Type Rechargeable, Sealed
Color Black
Product Type Remote Area Lighting System