missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PEG 40 Castor Oil, Spectrum™ Chemical
Description
Spectrum™ Chemical PEG 40 Castor Oil is a vegetable oil formed from the seeds of the castor oil plant. It is used to produce lubricants, hydraulic and brake fluids, pharmaceuticals, and cold resistant plastics. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
Specifications
Specifications
| CAS Max % | 1 |
| Acid Value | 2 |
| Quantity | 4 L |
| Grade | Reagent |
| Moisture | 0.03 |
| Packaging | Amber Glass Bottle |