missing translation for 'onlineSavingsMsg'
Learn More

PEG 40 Castor Oil, Spectrum™ Chemical

Catalog No. 18605001
Change view
Click to view available options
Quantity:
4 L
20 L
Packaging:
Amber Glass Bottle
Steel Can
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18605001 20 L Steel Can
18605002 4 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18605001 Supplier Spectrum Chemical Mfg Cor Supplier No. P240420LTBL
Get Up to $500 in Chemicals with Purchase
View all promotions

P2404, 61791-12-6

Spectrum™ Chemical PEG 40 Castor Oil is a vegetable oil formed from the seeds of the castor oil plant. It is used to produce lubricants, hydraulic and brake fluids, pharmaceuticals, and cold resistant plastics. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 61791-12-6

Specifications

Acid Value 2
CAS Max % 1
Quantity 20 L
Grade Reagent
Moisture 0.03
Packaging Steel Can