Learn More
Thermo Scientific™ PEEK™ Union for High-Pressure HPLC Connections, 0.020 in. through hole, 0.28µL swept volume
Ensure reliable high-pressure connections with biocompatible Thermo Scientific™ PEEK™ Unions, Tees and Crosses for High-Pressure HPLC Connections.
Supplier: Thermo Scientific™ UP P 704
Description
Minimize system dead volumes and ensure reliable high-pressure connections in your HPLC system with Thermo Scientific™ PEEK™ Unions, Tees and Crosses for High-Pressure HPLC Connections. The material is biocompatible for a wide range of applications.
- Absolute zero or low dead volume formats
- Biocompatible