Learn More
Cytiva PEEK Stop Plug
Enables connection of HiTrap columns to a syringe, laboratory pump or chromatography system. Cytiva PEEK Stop Plugs are used to seal the top of a chromatography column, e.g. HiTrap™ prepacked columns. Fingertight or standard 1/16 in. or standard 5/16 in. male stop plugs available.
$118.49
Specifications
| Type | Stop Plug |
|---|---|
| Material | PEEK |
| For Use With (Equipment) | Chromatography Columns |
Description
The inlet and outlet design of HiTrap columns eliminates the need for column connectors when used with ÄKTAdesign™ chromatography systems. Connection to a syringe, laboratory pump, or chromatography system such as FPLC™ System is also possible using these connectors.- Made of durable polyetheretherketone (PEEK)
- One fingertight stop plug is connected to each HiTrap column at delivery
- Available in narrow 1/16 in., standard 1/16 in. and standard 5/16 in. male stop plugs
Specifications
| Stop Plug | |
| PEEK | |
| Chromatography Columns |