Learn More
Cytiva PEEK Stop Plug
Catalog No. 361041175
Change view
Click to view available options
Connections:
Stop Plug 1/16 in. Male
Catalog No. Connections
361041175 Stop Plug 1/16 in. Male
1 options
Catalog No. 361041175 Supplier Cytiva Supplier No. 18111252
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Enables connection of HiTrap columns to a syringe, laboratory pump or chromatography system. Cytiva PEEK Stop Plugs are used to seal the top of a chromatography column, e.g. HiTrap™ prepacked columns. Fingertight or standard 1/16 in. or standard 5/16 in. male stop plugs available.
The inlet and outlet design of HiTrap columns eliminates the need for column connectors when used with ÄKTAdesign™ chromatography systems. Connection to a syringe, laboratory pump, or chromatography system such as FPLC™ System is also possible using these connectors.
  • Made of durable polyetheretherketone (PEEK)
  • One fingertight stop plug is connected to each HiTrap column at delivery
  • Available in narrow 1/16 in., standard 1/16 in. and standard 5/16 in. male stop plugs
Specifications
Type Stop Plug
Connections Stop Plug 1/16 in. Male
Quantity 6 Pk.
Material PEEK
For Use With (Equipment) Chromatography Columns