missing translation for 'onlineSavingsMsg'
Learn More

Cytiva PEEK Connector 1/16 in. Luer

Catalog No. 361041178
Change view
Click to view available options
Connections:
1/16 in. Tapered Male to Luer Female
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Connections
361041178 1/16 in. Tapered Male to Luer Female
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 361041178 Supplier Cytiva Supplier No. 18111251

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Set of two (2) polyetheretherketone 1/16 in. Male/Luer Female connectors for connection of a syringe to the top of a chromatography column in the ÄKTA Chromatography System.

ÄKTA Laboratory-scale Systems are liquid chromatography systems used for protein purification at research laboratory scale.
  • Tapered syringe ends
  • Polyetheretherketone (PEEK) connstruction
  • Connections: 1/16 in. male–Luer female

Specifications

Type Connector
Connections 1/16 in. Tapered Male to Luer Female
Quantity Set of 2
Material PEEK
Color Black
For Use With (Equipment) ÄKTA Laboratory-Scale Systems, Syinge