Learn More
Fisherbrand™ Pediatric Tube Adapter
Catalog No. 23900210
Change view
Click to view available options
Capacity:
0.5 to 1 mL
Catalog No. Capacity
23900210 0.5 to 1 mL
1 missing translation for 'options'
Catalog No. 23900210 missing translation for 'mfr' Fisherbrand™ missing translation for 'supplierNo' 7713068K
Accessory for use with centrifuges.
  • Refer to centrifuge manual for compatibility confirmation
Specifications
Type Insert
Tube Type Pediatric, Conical, Round Bottom, Microliter
Material Plastic
Capacity 0.5 to 1 mL
Autoclavable Not Autoclavable