missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Pectolyase Y-23 (A. Niger)

Catalog No. MP08320971
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP08320971 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP08320971 Supplier MP Biomedicals™ Supplier No. 08320971
Only null left

Highly purified enzyme from Aspergillus niger used for isolation of biologically active protoplasts from a wide spectrum of higher plants and tissues.

  • Contains two types of pectinase such as endopolygalacturonase and endo-pectin lyase in high activity
  • Included is a maceration stimulating factor which stimulates tissue maceration by both pectinases
  • One unit is defined as an increase in A235 by 1.0 in the reaction mixture per minute

Specifications

Purity >1000 units per gram (pectin lyase)
Content And Storage Store at 2 to 8°C
Format Powder
For Use With (Application) Isolation of biologically active protoplasts
Quantity 1 g