missing translation for 'onlineSavingsMsg'
Learn More

Pectin for use with Total Dietary Fiber Control Kit, TDF-C10, MilliporeSigma™ Supelco™

Catalog No. 111015764
Change view
Click to view available options
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
111015764 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015764 Supplier Merck Emd Millipore Supplier No. P75361G
Only null left

Pectin is a major component of primary cell walls of all land plants. It encompasses a range of galacturonic acid-rich polysaccharides, and finds applications in food, pharmaceuticals and other industries.

Food and Beverages and General Analytical

Specifications

Physical Form Lyophilized Powder
CAS 9000-69-5
MDL Number MFCD00081838
Quantity 1 g
Packaging Poly Bottle