missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pectin, MP Biomedicals™
$126.95 - $126.95
Chemical Identifiers
| CAS | 9000-69-5 |
|---|---|
| Molecular Formula | (C7H10O7)n |
| Synonym | Poly-D-galacturonic acid methyl ester |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN10258780
|
MP Biomedicals Inc
0210258780 |
100 g |
Each for $126.95
|
|
|||||
Chemical Identifiers
| 9000-69-5 | |
| (C7H10O7)n | |
| Poly-D-galacturonic acid methyl ester |
Specifications
| Beige to White | |
| 9000-69-5 | |
| Poly-D-galacturonic acid methyl ester | |
| 20,000 to 400,000 |
| Powder | |
| (C7H10O7)n | |
| 100 g | |
| Pectin |