missing translation for 'onlineSavingsMsg'
Learn More

Pectin, MP Biomedicals™

Catalog No. ICN10258780
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10258780 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN10258780 Supplier MP Biomedicals Inc Supplier No. 0210258780

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 9000-69-5
Molecular Formula (C7H10O7)n
Synonym Poly-D-galacturonic acid methyl ester

Specifications

Color Beige to White
Physical Form Powder
Quantity 100 g
Formula Weight 20,000 to 400,000
Chemical Name or Material Pectin