missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pectin, Citrus, USP, Spectrum™ Chemical
Description
Spectrum™ Chemical Pectin, Citrus, USP is used as a gelling agent and thickening agent. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
| CAS Max % | 1 |
| Quantity | 500 g |
| Grade | USP |
| Loss on Drying | 0.1 |
| Packaging | Poly Bottle |