missing translation for 'onlineSavingsMsg'
Learn More

Pectin, Citrus, USP, Spectrum™ Chemical

Numéro de catalogue. 18604997
Change view
Click to view available options
Quantity:
500 g
2.5 kg
20 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity Packaging
18604997 500 g Poly Bottle
18604998 2.5 kg Poly Pail
18604999 20 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. 18604997 Fournisseur Spectrum Chemical Mfg Cor Code fournisseur PE100500GM
Il en reste null

PE100, 9000-69-5

Spectrum™ Chemical Pectin, Citrus, USP is used as a gelling agent and thickening agent. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Identifiants chimiques

CAS 9000-69-5

Spécifications

CAS Max % 1
Quantity 500 g
Grade USP
Loss on Drying 0.1
Packaging Poly Bottle