missing translation for 'onlineSavingsMsg'
Learn More

Pectin Citrus

Catalog No. AAJ6102136
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6102136 500 g
AAJ6102122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6102136 Supplier Thermo Scientific Chemicals Supplier No. J6102136

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 9000-69-5

Pectin Citrus is promoted in dietary supplement form as an alternative cancer treatment. It can also been used to treat prostate cancer and melanoma. It can also be used as a gelling agent, filler, and stabilizer. It Supports celluar health, promotes immune function, supports cardiovascular health, supports healthy function of multiple organs, Safely chelates heavy metals and toxins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(RUO – Research Use Only.)  Promoted in dietary supplement form as an alternative cancer treatment, Pectin Citrus can also been used to treat prostate cancer and melanoma. It can also be used as a gelling agent, filler, and stabilizer. It Supports celluar health, promotes immune function, supports cardiovascular health, supports healthy function of multiple organs, Safely chelates heavy metals and toxins.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.

Chemical Identifiers

CAS 9000-69-5
MDL Number MFCD00081838
Synonym Poly-D-galacturonic acid methyl ester

Specifications

Color Brown to Yellow
Quantity 500 g
Merck Index 14,7063
Solubility Information It is soluble in water.
Physical Form Powder
Chemical Name or Material Pectin Citrus
EINECSNumber 232-553-0
RTECSNumber RX4280000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only