missing translation for 'onlineSavingsMsg'
Learn More

Pectin from Citrus, TCI America™

Catalog No. p-7134381
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P002425G 25 g
P0024500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P002425G Supplier TCI America Supplier No. P002425G
Only null left

Chemical Identifiers

CAS 9000-69-5
MDL Number MFCD00081838

Specifications

Quantity 25 g
Physical Form Crystalline Powder
Chemical Name or Material Pectin from Citrus