missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Pectin, pract.

CAS: 9000-69-5

Supplier:  Thermo Scientific Chemicals 416860010

 View more versions of this product

Catalog No. AC416860010


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Orange to Orange-Red
3.0 to 3.8 (1% soln.)
9000-69-5
MFCD00081838
1 kg
Practical
12% max. (105°C, 2 hrs)
2% Max. (>60 Mesh)
Pectin
Powder
acid-insoluble ash: < 1%
15, 7169
67 to 71% (degree of esterification)
Authentic
Plastic bottle
30.5 to 36.5

Chemical Identifiers

9000-69-5
MFCD00081838
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only