missing translation for 'onlineSavingsMsg'
Learn More

Peanut oil

CAS: 8002-03-7

Supplier:  Thermo Scientific Chemicals 416850025

 View more versions of this product

Catalog No. AC416850025


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Peanut oil
8002-03-7
2.5 L
283°C
Arachis oil, Groundnut oil
Saturated fatty acids (chain length <C16): 0.4% max.
Glass bottle
0.9150g/mL
Yellow
Viscous Liquid
MFCD00131805
14, 7059
Solubility in water: insoluble. Other solubilities: very slightly soluble in alcohol,miscible with light petroleum
Authentic
0.915

Chemical Identifiers

8002-03-7
MFCD00131805
Arachis oil, Groundnut oil
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 232-296-4

RTECSNumber : RX2830000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only