missing translation for 'onlineSavingsMsg'
Learn More
Learn More
International Enviroguard™ PE Sleeves
Description
Lightweight 18 in. protective arm sleeves protect against particulates and other debris with PE Coated Polypropylene. Secure elastic bands on each end help keep sleeves in place to help prevent skin exposure, even with repetitive movements. These disposable sleeves are lightweight, blue in color, and tear-resistant, so you're protected throughout your shift. Sleeve covers are often used to protect workers' wrists and forearms from exposure to hazardous solutions or chemicals. All sleeves are manufactured for a secure and comfortable fit using lightweight fabric and elastic closures.
- Secure elastic bands on each end to help prevent skin exposure by keeping sleeves in place
- 18 in. length offers excellent arm coverage
- Made of tear-resistant and durable PE-coated polypropylene
Specifications
Specifications
| Product Type | Controlled Environment Sleeve |
| Material | Polypropylene |
| Color | Blue |
| Sterility | Non-sterile |
| Disposable | Single-use |
| Closure Type | Elastic Ends |
| Seam Type | Seamless |
| Latex Free | Yes |
| Model | 2185 |
| Length (English) | 18 in. |
| Show More |