Papain Vial, Papain Dissociation System, A Vial Reconstituted with 5mL of EBSS or Equivalent Yields a Solution at 20units of papain/mL in 1mM L-cysteine with 0.5mM EDTA, Store at 2 deg. to 8 deg.C, Source: Papaya Latex, 1 Vial
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Papain Vial, Papain Dissociation System, A Vial Reconstituted with 5mL of EBSS or Equivalent Yields a Solution at 20units of papain/mL in 1mM L-cysteine with 0.5mM EDTA, Store at 2 deg. to 8 deg.C, Source: Papaya Latex, 1 Vial