missing translation for 'onlineSavingsMsg'
Learn More

PDI™ Castile Soap Towelette

Catalog No. 360994377
Change view
Click to view available options
Quantity:
1000/Cs.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
360994377 1000/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 360994377 Supplier PDI™ Supplier No. D41900
Only null left

Ideal for midstream clean-catch urine specimen procedures

  • Contains gentle, pH balanced cleansers
  • With 2% coconut oil

Specifications

Product Type Castile Soap Towelette
Composition Coconut oil 2%
For Use With (Application) Ideal for midstream clean-catch urine specimen procedures
Quantity 1000/Cs.