Learn More
SignalChem Biotech AMPK (A1/B1/G3), Active 50ug
Catalog No. 502573607
Encompass
Recombinant full-length human AMPK (combination of A1/B1/G3 subunits) was expressed by baculovirus in Sf9 insect cells using the N-terminal GST and C-terminal His tags.
Catalog No. 502573607 Supplier SignalChem Biotech Supplier No. P5610G50
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Recombinant full-length human AMPK (combination of A1/B1/G3 subunits) was expressed by baculovirus in Sf9 insect cells using the N-terminal GST and C-terminal His tags.