Learn More
ACROBiosystems Inc ACROBiosystems Mouse PD-L1 / B7-H1 Protein, Fc Tag
Catalog No. 501957259
Encompass
Recombinant Protein;1MG;Mouse PD-L1, Fc Tag (PD1-M5251) is expressed from human 293 cells (HEK293). It contains AA Phe 19 - Thr 238 (Accession # NP_068693).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 51.4 kDa. The protein migrates as 70-95 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Mouse PD-L1, Fc Tag (Cat. No. PD1-M5251) at 10 g/mL (100 L/well) can bind Biotinylated Mouse PD-1, Fc,Avitag (Cat. No. PD1-M82F4) with a linear range of 0.156-2.5 g/mL (QC tested).
Catalog No. 501957259 Supplier ACROBiosystems Inc Supplier No. PD1M52511MG
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Recombinant Protein;1MG;Mouse PD-L1, Fc Tag (PD1-M5251) is expressed from human 293 cells (HEK293). It contains AA Phe 19 - Thr 238 (Accession # NP_068693).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 51.4 kDa. The protein migrates as 70-95 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Mouse PD-L1, Fc Tag (Cat. No. PD1-M5251) at 10 g/mL (100 L/well) can bind Biotinylated Mouse PD-1, Fc,Avitag (Cat. No. PD1-M82F4) with a linear range of 0.156-2.5 g/mL (QC tested).