Learn More
Abcam Inc PD 173074, FGFR1 and FGFR3 inhibitor, 10MG
Catalog No. 502706888
Encompass
MW 523.7 Da, Purity >99%. Potent, ATP-competitive fibroblast growth factor receptor 1 (FGFR1) and FGFR3 inhibitor (IC values are 21.5 and 5 nM respectively). Shows little activity at EGFR, InsR, MEK and cPKC. Effectively inhibits angiogenesis in vivo. Also impairs tumor growth and inhibits self-renewal of stem cells.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 502706888 Supplier Abcam Inc Supplier No. AB14111710MG
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
MW 523.7 Da, Purity >99%. Potent, ATP-competitive fibroblast growth factor receptor 1 (FGFR1) and FGFR3 inhibitor (IC values are 21.5 and 5 nM respectively). Shows little activity at EGFR, InsR, MEK and cPKC. Effectively inhibits angiogenesis in vivo. Also impairs tumor growth and inhibits self-renewal of stem cells.
The product is subject to the following: Abcam Restricted Use Statement