Learn More
Research Products Corporation PCR Tube Mini-Freezer Box with Divider, 64 x 0.2ml Tube Capacity, 5 per Case
Catalog No. 501367533
Encompass
New Mini-Freezer box with insert designed exclusively for PCR tubes including 8 strip formats and individual tubes. Each box will accommodate up to 64 x 0.2ml tubes in an 8 x 8 array. Box features a one inch shallow depth to provide easy access to tubes. Outer box dimensions are 3" x 3" x 1" high and can be double stacked for storing in a standard freezer rack.
Catalog No. 501367533 Supplier Research Products Corporation Supplier No. 181057
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
New Mini-Freezer box with insert designed exclusively for PCR tubes including 8 strip formats and individual tubes. Each box will accommodate up to 64 x 0.2ml tubes in an 8 x 8 array. Box features a one inch shallow depth to provide easy access to tubes. Outer box dimensions are 3" x 3" x 1" high and can be double stacked for storing in a standard freezer rack.