missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One PCR Strip Tubes without Cap
Description
- Non-Pyrogenic
- Thin wall for optimal heat transfer
- Low evaporation rate in PCR
Specifications
Specifications
| Color | Red |
| Volume (Metric) | 0.2 mL |
| For Use With (Equipment) | PCR |
| Sterility | Non-sterile |
| Material | Polypropylene |
| Product Type | PCR Tube Strip |