Learn More
Promega PCR Nucleotide Mix, 25mM, 200microl DFS Item
PCR Nucleotide Mix is a premixed solution containing the sodium salts of dATP, dCTP, dGTP and dTTP, each at a concentration of 10mM or 25mM in water.
Supplier:  Promega U1431
Catalog No. PRU1431
Only null left
Description
Description
  • cGMP−Grade Solution of dNTPs Pretested in PCR
  • cGMP−manufactured for lot−to−lot product consistency
  • Equimolar amounts of each dNTP in water
  • Two concentrations available: 10mM and 25mM
Specifications
Specifications
-30C TO -10C
200microl
Product Suggestions
Product Suggestions