missing translation for 'onlineSavingsMsg'
Learn More

PCR Mycoplasma detection kit

Catalog No. AAJ66117AMJ
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ66117AMJ 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ66117AMJ Supplier Thermo Scientific Chemicals Supplier No. J66117AMJ
Only null left

Includes

Mycoplasma PCR Detection Kit; for 100 reactions.

The PCR Mycoplasma Detection Kit streamlines the process of identifying a mycoplasma outbreak in laboratory facilities.

Mycoplasma PCR Detection Kit has been used for screening of mycoplasma infection.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description

• Mycoplasma are some of the smallest living organisms, making them difficult to identify

• Mycoplasma outbreaks are common and can devastate cell and tissue research overnight

• Alternative testing methods can be slow and inaccurate

Applications

• Highly specific for mycoplasma DNA—little to no cross-reactivity

• Suitable for identification of mycoplasma in cell and tissue cultures

Specifications

Chemical Name or Material PCR Mycoplasma detection kit
Physical Form Liquid
Quantity 100 μL
Solubility Information Soluble in water.

RUO – Research Use Only