Learn More
Eppendorf™ PCR Foil
Description
- Effective adhesive sealing prevents evaporation loss during PCR
- Removal from the plate with no residue
- Easily pierced
- No sticking of the pipette tip (ideal for automated systems)
Specifications
Specifications
| Sterility | Sterile |
| No. per Pack | 100 Pk. |
| For Use With (Application) | Ideal for Automated Systems, PCR, RT PCR |
| For Use With (Equipment) | Ideal for Automated Systems Like PCR, RT PCR |
| Product Type | PCR Films and Foils |
| Adhesive | Self |
| Pierceable | Yes |
| Temperature Range (Metric) | 121°C |