Learn More
Eppendorf™ PCR Films and Foils
Description
- Effective adhesive sealing prevents evaporation loss during PCR
- Removal from the plate with no residue
- Sample can be visually monitored through the transparent PCR film
- PCR Foil: Easily pierced
- PCR Foil: No sticking of the pipette tip (ideal for automated systems)
Specifications
Specifications
| Sterility | Sterile |
| Autoclavable | Autoclavable |
| No. per Pack | 100 |
| For Use With (Application) | Ideal for Automated Systems Like PCR, RT PCR |
| Product Type | PCR Films and Foils |
| Adhesive | Self |
| Pierceable | Yes |
| Temperature Range (Metric) | 121°C |