missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Sulfate, Crystal, Reagent, ACS, 99%, Spectrum™ Chemical

Catalog No. 18600664
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
6 x 500 g
12 kg
25 kg
4 x 2.5 kg
50 kg
Packaging:
Poly Bottle
Poly Drum
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18600664 25 kg Poly Bottle
18600656 125 g Poly Bottle
18600657 500 g Poly Bottle
18600659 2.5 kg Poly Bottle
18600658 6 x 500 g Poly Bottle
18600660 4 x 2.5 kg Poly Bottle
18600661 12 kg Poly Bottle
18600662 50 kg Poly Drum
18600663 50 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 18600664 Supplier Spectrum Chemical Mfg Cor Supplier No. A124525KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions

missing translation for 'validMainframeMsg'

A1245, 7783-20-2, (NH4)2SO4

Spectrum™ Chemical Ammonium Sulfate, Crystal, Reagent, ACS is an inorganic salt that is very soluble in water Most often used as a soil fertilizer to release minute amounts of acid. It is also used in biochemistry as a precipitant to purify protein, used as a food additive and flame retardant. As an ACS grade Reagent, Spectrum Chemical manufactured compound is used as the quality standard against which other substances are grade and has met the toughest regulatory standards for quality and pureness.

Chemical Identifiers

CAS 7783-20-2
Molecular Formula H8N2O4S
Molecular Weight (g/mol) 132.13
MDL Number MFCD00003391
InChI Key BFNBIHQBYMNNAN-UHFFFAOYSA-N
IUPAC Name sulfuric acid diamine
SMILES N.N.OS(O)(=O)=O

Specifications

pH 5.0-6.0
CAS Max % 100%
Physical Form Crystalline Powder or Needles
Quantity 25 kg
Percent Purity 99%
Grade Reagent
Packaging Poly Bottle
Residue after Ignition 0.005%
Chemical Name or Material Ammonium Sulfate, Crystal