Learn More
Foxx Life Sciences Autofil Sterile Autoclavable Polycarbonate (PC) Media Storage Bottles with SECUREgrasp GL45 Cap, 1000mL, 24/CS
Supplier:  Foxx Life Sciences 1182RLS
Encompass
1182-RLS: Media Bottle, 1000 mL, PC, Sterile. Foxx Life Sciences' Polycarbonate Media Bottles have been designed specifically for Life Science researchers to store common solutions such as tissue culture media, serum, and buffers. Our bottles are designed to handle full vacuum, and can be used with the Autofil filtration system. Our PC bottles are highly transparent, shatterproof, and have good alcohol resistance. They are gamma-sterilized and rated to withstand five autoclave cycles (bottle only). The bottles are sterilized in the USA. Each Autofil bottle has features that improve the handling of the bottle, especially when the bottle is wet. The narrow bottle shoulder, combined with the molded finger grips, improves the ability to securely grip the bottle with gloved hands. Our patented, SECUREgrasp bottle cap enables easy handling and carrying. The innovative cap also allows for easy uncapping, tightening, and bottle stacking, and provides a large surface for labeling.
Catalog No. 501050047
Only null left