missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ PC Carboy
Description
- Conduct large-volume media and culture preparation with these 10 L FISHERBRAND PC, Carboy, particularly when visual inspection is required.
- This polycarbonate Clearboy is durable, lighter and safer than glass, autoclavable for sterile applications and ideal for refrigerated or frozen storage of aqueous solutions such as sterile water, aqueous reagents and media.
- Features:
- Available in 10 and 20 L sizes for a variety of biopharmaceutical applications
- Graduated in liters and gallons
- Molded of transparent, durable polycarbonate
- Customization options available for fluid transfer applications
- Autoclavable, graduated, leakproof
- Includes: Polypropylene closure and TPE gasket
Spécifications
Spécifications
| Autoclavable | Autoclavable |
| Capacity (Metric) | 20 L |
| Color | Natural (Translucent) |
| Disposable | Reusable |
| Material | Polycarbonate |
| Mouth Style | Wide |
| Product Type | Carboy |
| Sterility | Non-sterile |