Learn More
Stemcell Technologies Dulbecco's Phosphate Buffered Saline with 2% Fetal Bovine Serum, Size: 500 mL
Supplier: Stemcell Technologies Inc STM07905
Dulbecco's Phosphate Buffered Saline with 2% Fetal Bovine Serum is recommended for use as a buffer to dilute or wash nucleated cells.