Learn More
Patulin, 99%
CAS: 149-29-1 | C7H6O4 | 154.12 g/mol
Supplier: Thermo Scientific Chemicals 228870100
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Patulin | |
| 149-29-1 | |
| White | |
| 98.5% min. (HPLC) | |
| C7H6O4 | |
| 10 mg | |
| patulin, clavacin, expansin, clavatin, claviform, expansine, patuline, clairformin, claviformin, gigantin | |
| ZRWPUFFVAOMMNM-UHFFFAOYSA-N | |
| 4-hydroxy-4,6-dihydrofuro[3,2-c]pyran-2-one | |
| 4696 | |
| 154.12 | |
| Crystalline Powder or Crystals |
| 99% | |
| 108.0°C to 111.0°C | |
| Authentic | |
| Glass bottle | |
| MFCD00005858 | |
| 15, 7158 | |
| Solubility in water: soluble. Other solubilities: soluble in most common organic solvents,soluble in ethyl and amyl acetate,insoluble in petroleum ether | |
| C1C=C2C(=CC(=O)O2)C(O1)O | |
| 154.12 | |
| CHEBI:74926 | |
| 99% |
Chemical Identifiers
| 149-29-1 | |
| 154.12 | |
| ZRWPUFFVAOMMNM-UHFFFAOYSA-N | |
| 4696 | |
| 4-hydroxy-4,6-dihydrofuro[3,2-c]pyran-2-one |
| C7H6O4 | |
| MFCD00005858 | |
| patulin, clavacin, expansin, clavatin, claviform, expansine, patuline, clairformin, claviformin, gigantin | |
| CHEBI:74926 | |
| C1C=C2C(=CC(=O)O2)C(O1)O |
Safety and Handling
GHS H Statement:
Toxic in contact with skin.
Toxic if inhaled.
Toxic if swallowed.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 205-735-2
RTECSNumber : LV2625000
RUO – Research Use Only