missing translation for 'onlineSavingsMsg'
Learn More

BD Patient/Exam Room Sharps Collectors

Catalog No. 14826143
Change view
Click to view available options
Capacity (English):
3 gal.
Color:
Yellow
Design:
Counterbalanced Door
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English) Color Design
14826143 3 gal. Yellow Counterbalanced Door
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14826143 Supplier BD Supplier No. 300452

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Horizontal, side or vertical entry

  • Strong, durable polypropylene construction gives containers excellent impact and puncture-resistance
  • Suitable for incineration or autoclaving
  • Clear top and door provide visual indication of fill level
  • Safety transport handles keep hands away from collector body

Horizontal- and Side-Entry Collectors

  • Counterbalanced door opens automatically for single-handed disposal; stays closed to prevent overfilling

Order Info

Shipped nested; assembly of top to base required.

Specifications

Capacity (Metric) 10.3 L
Design Counterbalanced Door
Capacity (English) 3 gal.
Color Yellow
Disposable Single-use
For Use With (Application) Suitable for incineration or autoclaving
Material Polypropylene