Learn More
Mopec PathOdorizer Cleaner, Degreaser, and Odor Eliminating Solution
Description
- Eliminates offensive odors emitted from decomposed bodies within seconds
- Developed specially for use by medical examiners
- Can be used in every phase of work from the discovery scene to the morgue
- Removes odors from clothes, vehicles, body bags, refrigerators and other items
- Keeps the odor from permeating the lab environment
- Water soluble
- Odorless
- Colorless
- Biodegradable
- Non-flammable
- Non-corrosive
Specifications
Specifications
| Product Type | Air Freshener |
| Color-coded | No |
| Form | Clear Liquid |
| Scent | Unscented |
| Sterility | Sterile |
| Appearance | Colorless |
| Description | Deodorizer, Cleaner and Degreaser |