Learn More
StatLab PathFlow PiSmart Thermal Transfer Attached Lid Front Hinge Biopsy Cassettes
Description
- 0.9 mm pores
- Front hinge with lid attached
- Made from acetal polymer
- Specifically validated for the Pi Smart Thermal Transfer Cassette Printer
- Print face features four raised pips in the corners to protect printed surface during dewaxing, while enabling the surface to be cleared of wax
- Threaded
- 2000 per case
Specifications
Specifications
| Quantity | 2000 Cassettes/Cs. |
| Color | Yellow |
| Product Type | Biopsy Cassettes |
Made in USA